Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD Difco™ Dehydrated Culture Media: Letheen Agar

Catalog No. DF0680178 Shop All BD Diagnostic Systems Products
Change view
Click to view available options
Quantity:
500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
DF0680-17-8 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. DF0680-17-8 Supplier BD Supplier No. 268010
Add to Cart
Add to Cart
This item is not returnable. View return policy

For evaluating the bactericidal activity of quaternary ammonium compounds

Applications:

For evaluation, maintenance, selective enrichment, differentiation, isolation, cultivation, detection, propagation of bacteria

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage <30°C
Form Dehydrated
Media Type Letheen Agar
Product Type Agar
Formulation Letheen Agar 32g/L, Tryptone 5g/L, Proteose Peptone No. 3 10g/L, Yeast extract 2g/L, Sodium Chloride 5g/L, Sodium Bisulfite 0.1g/L, Agar 5g/L
For Use With (Application) For evaluating the bactericidal activity of quaternary Ammonium compounds
Packaging Type Bottle
Quantity 500 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.