Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD Difco™ Dehydrated Culture Media: Letheen Broth (AOAC)

Catalog No. df0681177 Shop All BD Diagnostic Systems Products
Change view
Click to view available options
Quantity:
500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
DF0681-17-7 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. DF0681-17-7 Supplier BD Supplier No. 268110
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

For determining the phenol coefficient of cationic surface-active materials

Applications:  For evaluation, maintenance, selective enrichment, differentiation, isolation, cultivation, detection, propagation of bacteria.

Letheen Broth contains beef extract and peptone which provide the carbon and nitrogen sources required for growth of a wide variety of organisms. Lecithin and polysorbate 80 are added to neutralize surface disinfectants. Lecithin is added to neutralize quaternary ammonium compounds and polysorbate 80 is incorporated to neutralize phenols, hexachlorophene, formalin and, with lecithin, ethanol. We do not use it for phenol coefficient determination, but rather for the neutralization of the most commonly encountered antimicrobials in antimicrobial testing of textile products. 

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance AOAC
Content And Storage 2°C to 8°C
Form Dehydrated
Media Type Letheen Broth
Product Type Microbiology Media
Formulation Contains: 10.0g/L Proteose Peptone No. 3; 5.0g/L Beef Extract; 5.0g/L Sodium Chloride; 5.0g/L Polysorbate 80; 0.7g/L Lecithin
For Use With (Application) Determination of Phenol coefficient of cationic surface-active materials
Packaging Type Bottle
Quantity 500 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.