Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD Difco™ Dehydrated Culture Media: m E Agar

Catalog No. DF0333179 Shop All BD Diagnostic Systems Products
Change view
Click to view available options
Quantity:
500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
DF0333-17-9 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. DF0333-17-9 Supplier BD Supplier No. 233320
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Used with nalidixic acid and triphenyltetrazolium chloride in isolating and differentiating enterococci from water by membrane filtration and in an in situ esculin test on Esculin Iron Agar

  • Formulation: Yeast extract 30g/L, Peptone 10g/L, Sodium Chloride 15g/L, Esculin 1g/L, Cycloheximide 0.05g/L, Sodium Azide 0.15g/L, Agar 15g/L

Certifications

ISO, 9000, EPA, SMWW

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance ISO, 9000, EPA, SMWW
Content And Storage <30°C
Form Dehydrated
Media Type mE Agar
Product Type Agar
Formulation Yeast extract 30g/L, Peptone 10g/L, Sodium Chloride 15g/L, Esculin 1g/L, Cycloheximide 0.05g/L, Sodium Azide 0.15g/L, Agar 15g/L
For Use With (Application) Isolation of Enterococcus faecalis 29212, Enterococcus faecalis 33186, Escherichia coli 25922
Packaging Type Bottle
Quantity 500 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.