Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD Difco™ Dehydrated Culture Media: m Endo Broth MF™

Catalog No. DF0749177 Shop All BD Diagnostic Systems Products
Change view
Click to view available options
Quantity:
500 g
Form:
Dehydrated
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Form
DF0749-17-7 500 g Dehydrated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. DF0749-17-7 Supplier BD Supplier No. 274930
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

For enumerating coliform organisms in water by membrane filtration

  • Formulation: Yeast extract 1.5g Casitone 5g/L, Thiopeptone 5g/L, Tryptose 10g/L, Lactose 12.5g/L, Sodium Desoxycholate 0.1g/L, Dipotassium Phosphate 4.375g/L, Monopotassium Phosphate 1.375g/L, Sodium Chloride 5g/L, Sodium Lauryl Sulfate 0.05g/L, Sodium Sulfite 2.1g/L

Certifications

COMPF, SMD, SMWW

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance COMPF, SMD, SMWW
Content And Storage <30°C
Form Dehydrated
Media Type m Endo Broth MF-Millipore
Product Type Microbiology Media
Formulation Yeast extract 1.5g/L, Casitone 5g/L, Thiopeptone 5g/L, Tryptose 10g/L, Lactose 12.5g/L, Sodium Desoxycholate 0.1g/L, Dipotassium Phosphate 4.375g/L, Monopotassium Phosphate 1.375g/L, Sodium Chloride 5g/L, Sodium Lauryl Sulfate 0.05g/L, Sodium Sulfite 2.1g/L, Basic Fuchsin 1.05g/L
For Use With (Application) Millipore™ filter, one step membrane filtration technique for the enumeration of Coliforms
Packaging Type Bottle
Quantity 500 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.