Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD Difco™ Dehydrated Culture Media: M9 Minimal Salts, 5x

Catalog No. DF048517 Shop All BD Diagnostic Systems Products
Change view
Click to view available options
Quantity:
500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
DF0485-17 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. DF0485-17 Supplier BD Supplier No. 248510
Add to Cart
Edge
Add to Cart
This item is not returnable. View return policy

For cultivation of recombinant strains of E. coli. BD Difco™ Dehydrated Culture Media: M9 Minimal Salts, 5x is part of a large selection of dehydrated media.

  • Dehydrated
  • Culture Media
  • M9 Minimal Salts
  • 5x 

Order Info

See our web site or the Fisher Scientific™ HealthCare catalog for product information.

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Refrigerate
Form Dehydrated
Product Type Prepared Microbiology Media
Formulation Disodium Phosphate (anhydrous) 33.9g/L, Monopotassium Phosphate 15g/L, Sodium Chloride 2.5g/L, Ammonium Chloride 5g/L
For Use With (Application) Cultivation of recombinant strains of Escherichia coli
Packaging Type Bottle
Quantity 500 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.