Learn More
Description
Order Info
See our web site or the Fisher Scientific™ HealthCare catalog for product information.
Certifications
COMPF, SMD
Specifications
Specifications
| Certifications/Compliance | COMPF, SMD |
| Content And Storage | Refrigerate |
| Form | Dehydrated |
| Media Type | Malonate Broth |
| Product Type | Microbiology Media |
| Formulation | Ammonium Sulfate 2g/L, Dipotassium Phosphate 0.6g/L, Monopotassium Phosphate 0.4g/L, Sodium Chloride 2g/L, Sodium Malonate 3g/L, Bromthymol blue 25mg/L |
| For Use With (Application) | Differentiation of Enterobacter and Escherichia on the basis of malonate utilization |
| Packaging Type | Bottle |
| Quantity | 500 g |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.