Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD Difco™ Dehydrated Culture Media: Malt Agar

Catalog No. DF0024173 Shop All BD Diagnostic Systems Products
Change view
Click to view available options
Quantity:
500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
DF0024-17-3 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. DF0024-17-3 Supplier BD Supplier No. 224200
Add to Cart
Add to Cart
This item is not returnable. View return policy

For detection and isolation of yeasts and molds

Order Info

See our web site or the Fisher Scientific™ HealthCare catalog for product information.

Certifications

ISO 9001:2008, AOAC, BAM

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance ISO 9001:2008, AOAC, BAM
Content And Storage Refrigerate
Form Dehydrated
Media Type Malt Agar
Product Type Agar
Formulation Malt extract 30g/L, Agar 15g/L
For Use With (Application) Dectection and isolation of Yeasts and molds
Packaging Type Bottle
Quantity 500 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.