Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD Difco™ Dehydrated Culture Media: Marine Broth 2216

Catalog No. DF0791174 Shop All BD Diagnostic Systems Products
Change view
Click to view available options
Quantity:
500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
DF0791-17-4 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. DF0791-17-4 Supplier BD Supplier No. 279110
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Cultivates heterotrophic marine bacteria. BD Difco™ Dehydrated Culture Media: Marine Broth 2216 has a shelf life of 1825 days.

  • Dehydrated
  • Culture Media
  • Marine Broth 2216
TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance CE
Content And Storage Refrigerate
Form Dehydrated
Media Type Marine Broth 2216
Product Type Microbiology Media
Formulation Peptone 5g/L, Yeast extract 1g/L, Ferric Citrate 0.1g/L, Sodium Chloride 19.45g/L, Magnesium Chloride 5.9g/L, Magnesium Sulfate 3.24g/L, Calcium Chloride 1.8g/L, Potassium Chloride 0.55g/L, Sodium Bicarbonate 0.16g/L, Potassium Bromide 0.08g/L, Strontium
For Use With (Application) Cultivation of heterotropic marine bacteria
Packaging Type Bottle
Quantity 500 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.