Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD Difco™ Dehydrated Culture Media: McClung Toabe Agar Base

Catalog No. DF0941173 Shop All BD Diagnostic Systems Products
Change view
Click to view available options
Quantity:
500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
DF0941-17-3 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. DF0941-17-3 Supplier BD Supplier No. 294110
Add to Cart
Add to Cart
This item is not returnable. View return policy

Use with Egg Yolk Enrichment 50% for detection and isolation of C. perfringens from foods based on the lecithinase reaction

Order Info

See our web site or the Fisher Scientific™ HealthCare catalog for product information.

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance CE
Content And Storage 2°C to 8°C
Form Dehydrated
Media Type McClung Toabe Agar Base
Product Type Agar
Formulation Proteose Peptone 40g/L, Dextrose 2g/L, Disodium Phosphate 5g/L, Monopotassium Phosphate 1g/L, Sodium Chloride 2g/L, Magnesium Sulfate 0.1g/L, Agar 25g/L
For Use With (Application) Egg Yolk enrichment 50% for isolating and detecting Clostridium perfringens in food based on the Lecithinase reaction
Packaging Type Bottle
Quantity 500 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.