Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD Difco™ Dehydrated Culture Media: MIL Medium

Catalog No. DF1804177 Shop All BD Diagnostic Systems Products
Change view
Click to view available options
Quantity:
500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
DF1804-17-7 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. DF1804-17-7 Supplier BD Supplier No. 218041
Add to Cart
Add to Cart
This item is not returnable. View return policy

Used for differentiating Enterobacteriaceae based on motility, lysine decarboxylation, lysine deamination and indole production

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage <30°C
Form Dehydrated
Media Type MIL Medium
Product Type Motility Test Medium
Formulation Peptone 10g/L, Pancreatic digest of Casein 10g/L, Yeast extract 3g/L, L-Lysine HCl 10g/L, Dextrose 1g/L, Ferric Ammonium Citrate 0.5g/L, Bromcresol Purple 0.02g/L, Agar 2g/L
For Use With (Application) Antimicrobial susceptibility testing and isolation of microorganisms
Packaging Type Bottle
Quantity 500 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.