Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD Difco™ Dehydrated Culture Media: MIO Medium

Catalog No. DF0735173 Shop All BD Diagnostic Systems Products
Change view
Click to view available options
Quantity:
500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
DF0735-17-3 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. DF0735-17-3 Supplier BD Supplier No. 273520
Add to Cart
Add to Cart
This item is not returnable. View return policy

Used to demonstrate motility, indole production and ornithine decarboxylase activity for the differentiation of Enterobacteriaceae

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance CE, BAM
Content And Storage Refrigerate
Form Dehydrated
Media Type MIO Medium
Product Type Motility Test Medium
Formulation Yeast extract 3g/L, Peptone 10g/L, Tryptone 10g/L, L-Ornithine HCl 5g/L, Dextrose 1g/L, Agar 2g/L, Bromcresol Purple 0.02g/L
For Use With (Application) Demonstrate motility, indole production and Ornithine decarboxylase activity for the differentiation of Enterobacteriaceae
Packaging Type Bottle
Quantity 500 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.