Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD Difco™ Dehydrated Culture Media: Mitis Salivarius Agar

Catalog No. DF0298172 Shop All BD Diagnostic Systems Products
Change view
Click to view available options
Quantity:
500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
DF0298-17-2 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. DF0298-17-2 Supplier BD Supplier No. 229810
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Use with Chapman Tellurite Solution 1% for selective isolation of S. mitis, S. salivarius and enterococci

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance CE
Content And Storage Refrigerate
Form Dehydrated
Media Type Mitis Salivarius Agar
Product Type Agar
Formulation Pancreatic digest of Casein 6g/L, Proteose Peptone 9g/L, Proteose Peptone 5g/L, Dextrose 1g/L, Saccharose 50g/L, Dipotassium Phosphate 4g/L, Trypan blue 75mg/L, Crystal Violet 0.8mg/L, Agar 15g/L
For Use With (Application) Used with Tellurite solution 1% in isolating Streptococcus mitis, S. salivarius and enterococci, particularly from grossly contaminated specimen
Packaging Type Bottle
Quantity 500 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.