Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD Difco™ Dehydrated Culture Media: MSRV Medium

Catalog No. D186817 Shop All BD Diagnostic Systems Products
Change view
Click to view available options
Quantity:
500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D186817 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. D186817 Supplier BD Supplier No. 218681
Add to Cart
Add to Cart
This item is not returnable. View return policy

For detecting motile Salmonella in feces and food products

  • Used with Novobiocin Antimicrobic Supplement

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance CCAM
Content And Storage 2°C to 30°C
Form Dehydrated
Media Type MSRV Medium
Product Type MSRV Medium
Formulation Tryptose 4.59g/L, Casein Hydrolysate (acid) 4.59g/L, Sodium Chloride 7.34g/L, Monopotassium Phosphate 1.47g/L, Magnesium Chloride (anhydrous) 10.9g/L, Malachite green Oxalate 37mg/L, Agar 2.7g/L
For Use With (Application) Novobiocin antimicrobic supplement
Packaging Type Bottle
Quantity 500 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.