Learn More
Description
Mueller Hinton Broth, not cation-adjusted, is a dehydrated culture medium that is not supplemented with calcium or magnesium ions. It is formulated to have a low thymine and thymidine content. It may be used for broth dilution antimicrobial susceptibility testing as long as the calcium and magnesium ion concentrations are adjusted according to CLSI standard M7.
- Acid hydrolysate (digest) of casein and beef extract supply amino acids and other nitrogenous substances, minerals, vitamins, carbon and other nutrients to support the growth of microorganisms
- Starch acts as a protective colloid against toxic substances in the medium; hydrolysis of the starch during autoclaving provides a small amount of dextrose, which is a source of energy
- Displays a cultural response to: Enterococcus faecalis 29212, Escherichia coli 25922, Pseudomonas aeruginosa 27853 and Staphylococcus aureus 29213
- pH (2.1% solution, 25°C): 7.3 ± 0.1
- Calcium: 2.9–5.9mg/L; Magnesium: 3.2–5.2mg/L
Specifications
Specifications
| Certifications/Compliance | CE, CLSI |
| Color | Light beige (with a few dark specks) |
| Content And Storage | Refrigerate |
| Form | Dehydrated Powder |
| Media Type | Mueller Hinton Broth |
| Product Type | Nutrient Broth |
| Formulation | Beef extract 2g/L, Acid digest of Casein 17.5g/L, Starch 1.5g/L |
| For Use With (Application) | Cultivation of wide variety of fastidious and non-fastidious microorganisms |
| Packaging Type | Pail |
| Quantity | 2 kg |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.