Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD Difco™ Dehydrated Culture Media: Mueller Hinton Medium

Catalog No. DF0252078 Shop All BD Diagnostic Systems Products
Change view
Click to view available options
Content And Storage:
2°C to 30°C
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Content And Storage
DF0252-07-8 2°C to 30°C
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. DF0252-07-8 Supplier BD Supplier No. 225220
Add to Cart
Add to Cart
This item is not returnable. View return policy

For antimicrobial susceptibility testing of microorganisms by agar diffusion methods

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance BAM, BS12, CCAM, CLSI, CMPH2, ISO, MCM9
Content And Storage 2°C to 30°C
Form Dehydrated
Media Type Mueller Hinton Medium
Product Type Mueller Hinton Medium
Formulation Beef extract 2g/L, Acid digest of Casein 17.5g/L, Starch 1.5g/L, Agar 17g/L
For Use With (Application) For antimicrobial disc diffusion susceptibility testing of common, rapidly growing bacteria by the Bauer-Kirby method,1-3 as standardized by the clinical and laboratory standards institute
Packaging Type Pail
Quantity 2000 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.