Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD Difco™ Dehydrated Culture Media: MYP Agar Mannitol Yolk Polymyxin B

Catalog No. DF0810171 Shop All BD Diagnostic Systems Products
Change view
Click to view available options
Quantity:
500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
DF0810-17-1 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. DF0810-17-1 Supplier BD Supplier No. 281010
Add to Cart
Edge
Add to Cart
This item is not returnable. View return policy

Used with Egg Yolk Enrichment 50% and Antimicrobic Vial P for presumptive identification of B. cereus in foods

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance ISO 9001:2008, AOAC, BAM, COMPF, ISO, USDA
Content And Storage Refrigerate
Form Dehydrated
Media Type MYP Agar
Product Type Agar
Formulation Beef extract 1g/L, Peptone 10g/L, D-Mannitol 10g/L, Sodium Chloride 10g/L, Phenol red 25mg/L, Agar 15g/L
For Use With (Application) Egg Yolk enrichment 50% and antimicrobic Vial P for enumerating Bacillus cereus from food
Packaging Type Bottle
Quantity 500 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.