Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD Difco™ Dehydrated Culture Media: OF Basal Medium

Catalog No. DF0688170 Shop All BD Diagnostic Systems Products
Change view
Click to view available options
Quantity:
500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
DF0688-17-0 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. DF0688-17-0 Supplier BD Supplier No. 268820
Add to Cart
Add to Cart
This item is not returnable. View return policy

Oxidation/Fermentation medium for differentiation and classification of gram-negative microorganisms

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance CE, BAM, CCAM
Content And Storage Refrigerate
Form Dehydrated
Media Type OF Basal Medium
Product Type OF Basal Medium
Formulation Pancreatic digest of Casein 2g/L, Sodium Chloride 5g/L, Dipotassium Phosphate 0.3g/L, Bromthymol blue 0.08g/L, Agar 2g/L
For Use With (Application) Determination of oxidative and fermentative metabolism of Carbohydrate by gram negative rods on the basis of Acid reaction in either the open or closed system
Packaging Type Bottle
Quantity 500 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.