Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD Difco™ Dehydrated Culture Media: OGYE Agar Base Oxytetracycline Glucose Yeast Extract

Catalog No. DF1811178 Shop All BD Diagnostic Systems Products
Change view
Click to view available options
Quantity:
500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
DF1811-17-8 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. DF1811-17-8 Supplier BD Supplier No. 218111
Add to Cart
Add to Cart

Use with Antimicrobic Vial Oxytetracycline for selective isolation of yeasts and molds in foods

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance CE, CCAM, COMPF, IDF, ISO
Content And Storage Refrigerate
Form Dehydrated
Media Type Oxytetracycline Glucose Yeast Extract Agar Base
Product Type Agar
Formulation Yeast extract 5g/L, Dextrose 20g/L, Agar 12g/L
For Use With (Application) For use with the antimicrobial agent, Oxytetracycline, in isolating and enumerating Yeasts and molds in food
Packaging Type Bottle
Quantity 500 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.