Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD Difco™ Dehydrated Culture Media: Oxford Medium Base

Catalog No. DF0225170 Shop All BD Diagnostic Systems Products
Change view
Click to view available options
Quantity:
500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
DF0225-17-0 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. DF0225-17-0 Supplier BD Supplier No. 222530
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Use with Modified Oxford Antimicrobic Supplement or Oxford Antimicrobic Supplement to isolate L. monocytogenes

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance AOAC, BAM, CCAM, COMPF, ISO, SMD, USDA
Content And Storage Refrigerate
Form Dehydrated
Media Type Oxford Medium Base
Product Type Oxford Medium Base
Formulation Pancreatic digest of Casein 8.9g/L, Proteose Peptone 4.4g/L, Yeast extract 4.4g/L, Tryptic digest of Beef heart 2.7g/L, Starch 0.9g/L, Sodium Chloride 4.4g/L, Esculin 1g/L, Ferric Ammonium Citrate 0.5g/L, Lithium Chloride 15g/L, Agar 15.3g/L
For Use With (Application) For isolating and differentiating Listeria monocytogenes
Packaging Type Bottle
Quantity 500 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.