Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD Difco™ Dehydrated Culture Media: PALCAM Medium Base

Catalog No. DF063617 Shop All BD Diagnostic Systems Products
Change view
Click to view available options
PALCAM Medium Base; 500g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
DF0636-17 PALCAM Medium Base; 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. DF0636-17 Supplier BD Supplier No. 263620
Add to Cart
Add to Cart
This item is not returnable. View return policy

Use with PALCAM Antimicrobic Supplement in isolating and cultivating Listeria from foods

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance BAM, CCAM, COMPF, ISO, SMD
Content And Storage <30°C
Form Dehydrated
Media Type PALCAM Medium Base
Product Type PALCAM Medium base
Formulation Columbia Blood Agar base 39g/L (Pancreatic digest of Casein 10g/L, Proteose Peptone No. 3 5g/L, Yeast extract 5g/L, Beef heart infusion from 500 g 3g/L, Corn Starch 1g/L, Sodium Chloride 5g/L, Agar 15g/L); Mannitol 10g/L; Dextrose 0.5g/L; Esculin 1g/L; Ferric Ammonium Citrate 0.5g/L; Lithium Chloride 15g/L; Phenol red 0.08g/L; Acriflavine HCl 5mg/L; Polymyxin B Sulfate 0.01g/L; Agar 2g/L
For Use With (Application) For isolating and cultivating Listeria, particularly from foods and Milk products
Packaging Type Bottle
Quantity 500 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.