Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD Difco™ Dehydrated Culture Media: Pantothenate Assay Medium

Catalog No. DF0604153 Shop All BD Diagnostic Systems Products
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
DF0604-15-3 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. DF0604-15-3 Supplier BD Supplier No. 260410
Add to Cart
Add to Cart
This item is not returnable. View return policy

Microbiological assay of pantothenate

Order Info

See our web site or the Fisher Scientific™ HealthCare catalog for product information.

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage 2°C to 8°C
Form Dehydrated
Media Type Pantothenate Assay Medium
Product Type Pantothenate Assay Medium
Formulation Vitamin assay Casamino Acids 10g/L, Dextrose 40g/L, Sodium Acetate 20g/L, L-Cystine 0.4g/L, DL-Tryptophan 0.2g/L, Adenine Sulfate 20mg/L, Guanine Hydrochloride 20mg/L, Uracil 20mg/L, Thiamine Hydrochloride 200μg/L, Riboflavin 400
For Use With (Application) Microbiological assay of Pantothenate
Packaging Type Bottle
Quantity 100 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.