Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD Difco™ Dehydrated Culture Media: Phenylalanine Agar

Catalog No. DF0745171 Shop All BD Diagnostic Systems Products
Change view
Click to view available options
Quantity:
500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
DF0745-17-1 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. DF0745-17-1 Supplier BD Supplier No. 274520
Add to Cart
Add to Cart
This item is not returnable. View return policy

Differentiation of Proteus and Providencia from other Enterobacteriaceae based on deamination of phenylalanine

Order Info

See our web site or the Fisher Scientific™ HealthCare catalog for product information.

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance BAM
Content And Storage Refrigerate
Form Dehydrated
Media Type Phenylalanine Agar
Product Type Agar
Formulation DL-Phenylalanine 2g/L, Yeast extract 3g/L, Sodium Chloride 5g/L, Dipotassium Phosphate 1g/L, Agar 12g/L
For Use With (Application) For differentiation of enteric Bacilli on the basis of their ability to produce Phenylpyruvic Acid by oxidative deamination
Packaging Type Bottle
Quantity 500 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.