Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD Difco™ Dehydrated Culture Media: Pseudomonas Isolation Agar

Catalog No. DF0927171 Shop All BD Diagnostic Systems Products
Change view
Click to view available options
Quantity:
500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
DF0927-17-1 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. DF0927-17-1 Supplier BD Supplier No. 292710
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Used with Glycerol for selective isolation and differentiation of P. aeruginosa based on pigment formation

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage <30°C
Form Dehydrated
Media Type Pseudomonas Isolation Agar
Product Type Agar
Formulation Peptone 20g/L, Magnesium Chloride 1.4g/L, Potassium Sulfate 10g/L, Irgasan™ 25mg/L, Agar 13.6g/L
For Use With (Application) Used with Glycerol for selective isolation and differentiation of P. aeruginosa based on pigment formation
Packaging Type Bottle
Quantity 500 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.