Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD Difco™ Dehydrated Culture Media: Reinforced Clostridial Medium

Catalog No. DF1808173 Shop All BD Diagnostic Systems Products
Change view
Click to view available options
Quantity:
500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
DF1808-17-3 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. DF1808-17-3 Supplier BD Supplier No. 218081
Add to Cart
Edge
Add to Cart
This item is not returnable. View return policy

For cultivating and enumerating clostridia, other anaerobes, and other species of bacteria from foods and clinical specimens

  • Formulation: Peptone 10g/L, Beef extract 10g/L, Yeast extract 3g/L, Dextrose 5g/L, Sodium Chloride 5g/L, Soluble Starch 1g/L, Cysteine HCl 0.5g Sodium Acetate 3g Agar 0.5g/L

Order Info

Prices are for units of one each unless otherwise specified.

Certifications

CCAM, EP, JP, USP

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance CCAM, EP, JP, USP
Content And Storage <30°C
Form Dehydrated
Media Type Reinforced Clostridial Medium
Product Type Reinforced Clostridial Medium
Formulation Peptone 10g/L, Beef extract 10g/L, Yeast extract 3g/L, Dextrose 5g/L, Sodium Chloride 5g/L, Soluble Starch 1g/L, Cysteine HCl 0.5g Sodium Acetate 3g Agar 0.5g/L
For Use With (Application) Cultivation and enumeration of anaerobes, particularly Clostridia from foods and clinical specimens
Packaging Type Bottle
Quantity 500 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.