Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD Difco™ Dehydrated Culture Media: Rogosa Selective Lactobacillus (SL) Agar

Catalog No. DF0480170 Shop All BD Diagnostic Systems Products
Change view
Click to view available options
Quantity:
500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
DF0480-17-0 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. DF0480-17-0 Supplier BD Supplier No. 248020
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

For cultivating oral, vaginal and fecal lactobacilli

  • Tryptone 10g/L, Yeast extract 5g/L, Dextrose 10g/L, Arabinose 5g/L, Saccharose 5g/L, Sodium Acetate 15g/L, Ammonium Citrate 2g/L, Monopotassium Phosphate 6g/L, Magnesium Sulfate 0.57g Manganese Sulfate 0.12g Ferrous Sulfate 0.03g/L, Polysorbate 80 1g/L

Order Info

Prices are for units of one each unless otherwise specified.

Certifications

COMPF

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance COMPF
Content And Storage 2°C to 8°C
Form Dehydrated
Media Type Rogosa Selective Lactobacillus Agar
Product Type Agar
Formulation Tryptone 10g/L, Yeast extract 5g/L, Dextrose 10g/L, Arabinose 5g/L, Saccharose 5g/L, Sodium Acetate 15g/L, Ammonium Citrate 2g/L, Monopotassium Phosphate 6g/L, Magnesium Sulfate 0.57g Manganese Sulfate 0.12g Ferrous Sulfate 0.03g/L, Polysorbate 80 1g/L
For Use With (Application) Selective cultivation of oral, vaginal and fecal lactobacilli
Packaging Type Bottle
Quantity 500 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.