Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD Difco™ Dehydrated Culture Media: Sabouraud Agar, Modified (Dextrose Agar Emmons)

Catalog No. DF0747179 Shop All BD Diagnostic Systems Products
Change view
Click to view available options
Quantity:
500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
DF0747-17-9 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. DF0747-17-9 Supplier BD Supplier No. 274720
Add to Cart
Add to Cart
This item is not returnable. View return policy

For cultivation of pathogenic fungi

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance SMWW
Content And Storage <30°C
Form Dehydrated
Media Type Sabouraud Agar Modified (Dextrose Agar Emmons)
Product Type Agar
Formulation Enzymatic digest of Casein 10g/L, Dextrose 20g/L, Agar 20g/L
For Use With (Application) Cultivation of dermatophytes and other pathogenic and nonpathogenic fungi from clinical and nonclinical specimens
Packaging Type Bottle
Quantity 500 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.