Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD Difco™ Dehydrated Culture Media: Sabouraud Brain Heart Infusion Agar Base

Catalog No. DF0797178 Shop All BD Diagnostic Systems Products
Change view
Click to view available options
Quantity:
500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
DF0797-17-8 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. DF0797-17-8 Supplier BD Supplier No. 279720
Add to Cart
Add to Cart
This item is not returnable. View return policy

Use with chloromycetin and blood (optional) in cultivation and isolation of pathogenic fungi

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage <30°C
Form Dehydrated
Media Type SABHI Agar Base
Product Type Agar
Formulation Brain heart digest 9.25g/L, Proteose Peptone 5g/L, Enzymatic digest of Casein 5g/L, Dextrose 21g/L, Sodium Chloride 2.5g/L, Disodium Phosphate 1.25g/L, Agar 15g/L
For Use With (Application) Cultivation of dermatophytes and other pathogenic and nonpathogenic fungi from clinical and nonclinical specimens
Packaging Type Bottle
Quantity 500 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.