Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD Difco™ Dehydrated Culture Media: Select APS™ Super Broth

Catalog No. B4312485 Shop All BD Diagnostic Systems Products
Change view
Click to view available options
Quantity:
500 g
10 kg
Packaging Type:
Bottle
Pail
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging Type
B4312485 500 g Bottle
B4312486 10 kg Pail
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. B4312485 Supplier BD Supplier No. 212485
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

To grow Escherichia coli to a high cell density

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance CE
Content And Storage Refrigerate
Form Dehydrated
Media Type Super Broth Base, Select APSI (Alternative Protein Source)
Product Type Nutrient Broth
Formulation Soy Hydrolysate 12g/L, Yeast extract 24g/L, Dipotassium Phosphate 11.4g/L, Monopotassium Phosphate 1.7g/L
For Use With (Application) Molecular Genetics medium used to grow Escherichia coli to a high cell density
Packaging Type Bottle
Quantity 500 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.