Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD Difco™ Dehydrated Culture Media: Selenite Cystine Broth

Catalog No. DF0687171 Shop All BD Diagnostic Systems Products
Change view
Click to view available options
Quantity:
500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
DF0687-17-1 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. DF0687-17-1 Supplier BD Supplier No. 268740
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

For selective enrichment of Salmonella from food and water

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance ISO, 9000, AOAC, BAM, CCAM, COMPF, ISO, SMD, SMWW
Content And Storage <30°C
Form Dehydrated
Media Type Selenite Cystine Broth
Product Type Microbiology Media
Formulation Pancreatic digest of Casein 5g/L, Lactose 4g/L, Sodium Phosphate 10g/L, Sodium Selenite 4g/L, L-Cystine 0.01g/L
For Use With (Application) Isolation of Salmonella enterica subspecies enterica serotype Typhimurium, Shigella sonnei
Packaging Type Bottle
Quantity 500 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.