Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD Difco™ Dehydrated Culture Media: Seven H11 Agar Base

Catalog No. L12203 Shop All BD Diagnostic Systems Products
Change view
Click to view available options
Quantity:
500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
L12203 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. L12203 Supplier BD Supplier No. 212203
Add to Cart
Add to Cart
This item is not returnable. View return policy

For isolation of Mycobacterium tuberculosis

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance ISO/DIS, 11014
Content And Storage 25°C
Form Dehydrated
Media Type Seven H11 Agar Base
Product Type Agar
Formulation Pancreatic digest of Casein 1g/L, Monosodium Glutamate 0.5g/L, Sodium Citrate 0.4g/L, Pyridoxine 1mg/L, Biotin 0.5mg/L, Ferric Ammonium Citrate 0.04g/L, Ammonium Sulfate 0.5g/L, Disodium Phosphate 1.5g/L, Monopotassium Phosphate 1.5g/L, Magnesium Sulfate
For Use With (Application) Isolation of Mycobacterium tuberculosis
Packaging Type Bottle
Quantity 500 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.