Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD Difco™ Dehydrated Culture Media: SFP Agar Base

Catalog No. DF0811170 Shop All BD Diagnostic Systems Products
Change view
Click to view available options
Quantity:
500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
DF0811-17-0 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. DF0811-17-0 Supplier BD Supplier No. 281110
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Use with Egg Yolk Enrichment 50%, Antimicrobic Vial P and Antimicrobic Vial K for detection and enumeration of C. perfringens in foods

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance AOAC, BAM, COMPF, ISO
Content And Storage <30°C
Form Dehydrated
Media Type Shahidi Ferguson Perfringens Agar Base
Product Type Agar
Formulation Yeast extract 5g/L, Proteose Peptone No. 3 7.5g/L, Pancreatic digest of Casein 7.5g/L, Soytone 5g/L, Ferric Ammonium Citrate 1g/L, Sodium Bisulfite 1g/L, Agar 20g/L
For Use With (Application) Detection and enumerating Clostridium perfringens in foods
Packaging Type Bottle
Quantity 500 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.