Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD Difco™ Dehydrated Culture Media: Spirit Blue Agar

Catalog No. DF0950171 Shop All BD Diagnostic Systems Products
Change view
Click to view available options
Quantity:
500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
DF0950-17-1 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. DF0950-17-1 Supplier BD Supplier No. 295020
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Used with Lipase Reagent for enumeration and detection of lipolytic microorganisms

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance ISO/DIS, 11014, SMD
Content And Storage 25°C
Form Dehydrated
Media Type Spirit Blue Agar
Product Type Agar
Formulation Pancreatic digest of Casein 10g/L, Yeast extract 5g/L, Agar 20g/L, Spirit blue 0.15g/L
For Use With (Application) Isolation, selective enrichment, differentiation, enumeration, estimation, maintenance, cultivation of bacteria
Packaging Type Bottle
Quantity 500 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.