Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD Difco™ Dehydrated Culture Media: Thermoacidurans Agar

Catalog No. DF0303175 Shop All BD Diagnostic Systems Products
Change view
Click to view available options
Certifications/Compliance:
CCAM, COMPF
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Certifications/Compliance
DF0303-17-5 CCAM, COMPF
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. DF0303-17-5 Supplier BD Supplier No. 230310
Add to Cart
Add to Cart
This item is not returnable. View return policy

Isolation and cultivation of B. thermoacidurans from foods

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance CCAM, COMPF
Content And Storage <30°C
Form Dehydrated
Media Type Thermoacidurans Agar
Product Type Agar
Formulation Yeast extract 5g/L, Proteose Peptone 5g/L, Dextrose 5g/L, Dipotassium Phosphate 4g/L, Agar 20g/L
For Use With (Application) Isolation of pathogenic Neisseria from specimens containing mixed flora of bacteria and fungi
Packaging Type Bottle
Quantity 500 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.