Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD Difco™ Dehydrated Culture Media: Thioglycollate Medium without Indicator

Catalog No. DF0430171 Shop All BD Diagnostic Systems Products
Change view
Click to view available options
Quantity:
500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
DF0430-17-1 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. DF0430-17-1 Supplier BD Supplier No. 243010
Add to Cart
Add to Cart
This item is not returnable. View return policy

For detection of microorganisms in normally sterile materials, especially those containing mercurial preservatives

Fermentation studies

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance ISO/DIS, 11014, USP
Content And Storage <30°C
Form Dehydrated
Media Type Thioglycollate Medium without Indicator
Product Type Microbiology Media
Formulation Pancreatic digest of Casein 15g/L, Yeast extract 5g/L, Dextrose 5g/L, Sodium Chloride 2.5g/L, L-Cystine 0.25g/L, Sodium Thioglycollate 0.5g/L, Agar 0.75g/L
For Use With (Application) Isolation of obligate anaerobes
Packaging Type Bottle
Quantity 500 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.