Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD Difco™ Dehydrated Culture Media: Tomato Juice Broth

Catalog No. DF0517088 Shop All BD Diagnostic Systems Products
Change view
Click to view available options
Quantity:
10,000 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
DF0517-08-8 10,000 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. DF0517-08-8 Supplier BD Supplier No. 251710

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

For cultivation of yeasts and other aciduric microorganisms

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage 25°C
Form Dehydrated
Media Type Tomato Juice Broth
Product Type Microbiology Media
Formulation Tomato Juice (from 400mL) 20g/L, Yeast extract 10g/L, Dextrose 10g/L, Dipotassium Phosphate 0.5g/L, Monopotassium Phosphate 0.5g/L, Magnesium Sulfate 0.1g/L, Sodium Chloride 0.01g/L, Ferrous Sulfat 0.01g/L, Manganese Sulfate 0.01g/L
For Use With (Application) Isolation of Lactobacillus species
Packaging Type Pail
Quantity 10,000 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.