Learn More
BD Difco™ Dehydrated Culture Media: Tryptone Glucose Extract Agar
Shop All BD Diagnostic Systems ProductsDescription
Cultivation of bacteria in water, milk and other dairy products
Certifications
AOAC, COMPF
Specifications
Specifications
| Certifications/Compliance | AOAC, COMPF |
| Content And Storage | <30°C |
| Form | Dehydrated |
| Media Type | Tryptone Glucose Extract Agar |
| Product Type | Agar |
| Formulation | Beef extract 3g/L, Tryptone 5g/L, Dextrose (Glucose) 1g/L, Agar 15g/L |
| For Use With (Application) | Cultivation of a wide variety of fastidious microorganisms |
| Packaging Type | Bottle |
| Quantity | 500 g |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.