Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD Difco™ Dehydrated Culture Media: Violet Red Bile Glucose Agar

Catalog No. D186617 Shop All BD Diagnostic Systems Products
Change view
Click to view available options
Quantity:
500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D186617 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. D186617 Supplier BD Supplier No. 218661
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

For detecting and enumerating Enterobacteriaceae in food and dairy products

  • Formulation: Yeast extract 3g/L, Pancreatic digest of Gelatin 7g/L, Bile Salts No. 3 1.5g/L, Glucose 10g/L, Sodium Chloride 5g/L, Neutral red 0.03g/L, Crystal Violet 2g/L, Agar 15g/L

Order Info

Prices are for units of one each unless otherwise specified

Certifications

CCAM, COMPF, EP, ISO, JP, USP

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance CCAM, COMPF, EP, ISO, JP, USP
Color Violet Red
Content And Storage 2°C to 30°C
Form Dehydrated
Media Type Violet Red Bile Glucose Agar
Product Type Agar
Formulation Yeast extract 3g/L, Pancreatic digest of Gelatin 7g/L, Bile Salts No. 3 1.5g/L, Glucose 10g/L, Sodium Chloride 5g/L, Neutral red 0.03g/L, Crystal Violet 2g/L, Agar 15g/L
For Use With (Application) For detecting and enumerating Enterobacteriaceae in food and dairy products
Packaging Type Bottle
Quantity 500 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.