Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD Difco™ Dehydrated Culture Media: Wilkins-Chalgren Agar

Catalog No. DF1805176 Shop All BD Diagnostic Systems Products
Change view
Click to view available options
Quantity:
500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
DF1805-17-6 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. DF1805-17-6 Supplier BD Supplier No. 218051
Add to Cart
Add to Cart
This item is not returnable. View return policy

For susceptibility testing of anaerobes and for isolating and cultivating anaerobes

  • Formulation: Pancreatic digest of Casein 10g/L, Peptone 10g/L, Yeast extract 5g/L, Dextrose 1g/L, Sodium Chloride 5g/L, L-Arginine 1g/L, Sodium Pyruvate 1g/L, Hemin 5mg/L, Vitamin K1 0.5mg/L, Agar 15g/L

Order Info

Prices are for units of one each unless otherwise specified.

Certifications

ISO/DIS, 11014CCAM

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance ISO/DIS, 11014CCAM
Content And Storage <30°C
Form Dehydrated
Media Type Wilkins-Chalgren Agar
Product Type Agar
Formulation Pancreatic digest of Casein 10g/L, Peptone 10g/L, Yeast extract 5g/L, Dextrose 1g/L, Sodium Chloride 5g/L, L-Arginine 1g/L, Sodium Pyruvate 1g/L, Hemin 5mg/L, Vitamin K1 0.5mg/L, Agar 15g/L
For Use With (Application) Susceptibility testing of anaerobes and for isolating and cultivating anaerobes
Packaging Type Bottle
Quantity 500 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.