Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD Difco™ Dehydrated Culture Media: Wort Agar

Catalog No. DF0111177 Shop All BD Diagnostic Systems Products
Change view
Click to view available options
Quantity:
500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
DF0111-17-7 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. DF0111-17-7 Supplier BD Supplier No. 211671
Add to Cart
Add to Cart
This item is not returnable. View return policy

For cultivation and enumeration of yeasts

  • Formulation: Malt extract 15g/L, Peptone 0.78g/L, Maltose 12.75g/L, Dextrin 2.75g/L, Glycerol 2.35g/L, Dipotassium Phosphate 1g/L, Ammonium Chloride 1g/L, Agar 15g/L

Order Info

Prices are for units of one each unless otherwise specified.

Certifications

ISO/DIS, 11014

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance ISO/DIS, 11014
Content And Storage <30°C
Form Dehydrated
Media Type Wort Agar
Product Type Agar
Formulation Malt extract 15g/L, Peptone 0.78g/L, Maltose 12.75g/L, Dextrin 2.75g/L, Glycerol 2.35g/L, Dipotassium Phosphate 1g/L, Ammonium Chloride 1g/L, Agar 15g/L
For Use With (Application) For cultivation and enumeration of yeasts
Packaging Type Bottle
Quantity 500 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.