Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD Difco™ Dehydrated Culture Media: XLT4 Agar Base

Catalog No. DF0234179 Shop All BD Diagnostic Systems Products
Change view
Click to view available options
Quantity:
500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
DF0234-17-9 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. DF0234-17-9 Supplier BD Supplier No. 223420

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Used with XLT4 Agar Supplement in isolating non-typhi Salmonella

  • Formulation: Proteose Peptone No. 3 1.6g/L, Yeast extract 3g/L, L-Lysine 5g/L, Xylose 3.75g/L, Lactose 7.5g/L, Saccharose 7.5g/L, Ferric Ammonium Citrate 0.8g/L, Sodium Thiosulfate 6.8g/L, Sodium Chloride 5g/L, Agar 18g/L, Phenol red 0.08g/L

Order Info

Prices are for units of one each unless otherwise specified.

Certifications

ISO, 9000, USDA

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance ISO, 9000, USDA
Content And Storage <30°C
Form Dehydrated
Media Type XLT4 Agar Base
Product Type Agar
Formulation Proteose Peptone No. 3 1.6g/L, Yeast extract 3g/L, L-Lysine 5g/L, Xylose 3.75g/L, Lactose 7.5g/L, Saccharose 7.5g/L, Ferric Ammonium Citrate 0.8g/L, Sodium Thiosulfate 6.8g/L, Sodium Chloride 5g/L, Agar 18g/L, Phenol red 0.08g/L
For Use With (Application) Isolation of Enterococcus faecalis 29212, Escherichia coli 25922, Proteus mirabilis 25933, Salmonella enterica subspecies enterica serotype Typhimurium 14028, Staphylococcus aureus 25923
Packaging Type Bottle
Quantity 500 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.