Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD Difco™ Dehydrated Culture Media: Yeast Nitrogen Base

Catalog No. DF0392159 Shop All BD Diagnostic Systems Products
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
DF0392-15-9 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. DF0392-15-9 Supplier BD Supplier No. 239210
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Wickerham formula, classification of yeasts based on nitrogen assimilation. BD Difco™ Dehydrated Culture Media: Yeast Nitrogen Base is part of a large selection of dehydrated media.

  • Wickerham formula
  • Dehydrated
  • Culture Media
  • Yeast Nitrogen Base

Order Info

Prices are for units of one each unless otherwise specified.

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage <30°C
Form Dehydrated
Media Type Yeast Nitrogen Base
Product Type Microbiology Media
Formulation Nitrogen Source: Ammonium Sulfate 5g/L, Amino Acids: L-Histidine Monohydrochloride 10mg/L, LD-Methionine 20mg/L, LD-Tryptophan 20mg/L, Vitamins: Biotin 2μg/L, Calcium Pantothenate 400μg/L, Folic Acid 2μ
For Use With (Application) For classifying Yeasts based on colonial characteristics and cell morphology
Packaging Type Bottle
Quantity 100 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.