Learn More
Description
D-Glucose is a simple sugar that circulates in the blood of animals as blood sugar, and is made in plants during photosynthesis. It is an important source of power for cellular respiration. Glucose is stored as as a starch in plants and as glycogen in animals.
- CAS: 50-99-7
- EC No.: 200-075-1
- Chemical formula: C6H12O6
- Molar mass: 180.16g/mol
- Synonym: Blood Sugar
Specifications
Specifications
| Certifications/Compliance | ISO/DIS, 11014 |
| Color | Whitish |
| Content And Storage | <30°C |
| Additive | Dextrose Glucose |
| Form | Anhydrous Solid |
| For Use With (Application) | Microbiological culture media |
| Packaging Type | Pail |
| Quantity | 10 kg |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.