Learn More
Description
Listeria monocytogenes is a widespread problem in public health and the food industries.
- Lyophilized, polyclonal rabbit antisera containing approximately 0.1% sodium azide as a preservative
- Listeria O Antisera Types 1 and 4 are specific for the respective serotypes of L. monocytogenes while Listeria O Antiserum Poly contains agglutinins for L. monocytogenes serotypes 1 and 4
For in vitro Diagnostic Use.
Certifications
CE marked
Specifications
Specifications
| Certifications/Compliance | CE Marked; For in vitro diagnostic use |
| Content And Storage | Refrigerate, store at 2°C to 8°C |
| Product Type | Listeria O Antiserum Type 1 |
| For Use With (Application) | Used as positive control in identification of Listeria monocytogenes in the slide agglutination test |
| Quantity | 1 mL |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.