Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD BD™ Difco™ R2A Agar

Catalog No. DF1826171 Shop All BD Diagnostic Systems Products
Change view
Click to view available options
Quantity:
500 g
2 kg
Packaging Type:
Bottle
Pail
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging Type
DF1826-17-1 500 g Bottle
DF1826-07-3 2 kg Pail
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. DF1826-17-1 Supplier BD Supplier No. 218263
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Intended for use with stressed and chlorine-tolerant bacteria. BD™ Difco™ R2A Agar is a dehydrated, low-nutrient medium for enumerating and cultivating compromised bacteria and other heterotrophic organisms. It was developed for bacteriological plate counts of treated potable water.

R2A Agar was developed for bacteriological plate counts of treated potable water. A low nutrient medium, such as R2A Agar, in combination with a lower incubation temperature and longer incubation time stimulates the growth of stressed and chlorine-tolerant bacteria.

Nutritionally-rich media, such as Plate Count Agar, support the growth of fast-growing bacteria but may suppress slow growing or stressed bacteria found in treated water. When compared with nutritionally rich media, R2A Agar has been reported to improve the recovery of stressed and chlorine-tolerant bacteria from drinking water systems.

  • Recommended in standard methods for pour plate, spread plate and membrane filter methods for heterotrophic plate counts.
  • Intended for use only with compromised bacteria found in treated potable water.
  • Yeast extract provides a source of trace elements and vitamins.
  • Peptone and casamino acids provide nitrogen, vitamins, amino acids, carbon and minerals.
  • Dextrose serves as a carbon source.
  • Soluble starch aids in the recovery of injured organisms by absorbing toxic metabolic by-products.
  • Sodium pyruvate increases the recovery of stressed cells.
  • Potassium phosphate is used to balance the pH and provide phosphate.
  • Magnesium sulfate is a source of divalent cations and sulfate.
  • Agar is the solidifying agent.

Order Info

Prices are for units of one each unless otherwise specified.

Certifications

COMPF, EPA, SMWW

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance COMPF, EPA, SMWW
Color Light beige
Form Dehydrated (free-flowing, homogenous)
Media Type R2A Agar
Product Type Agar
Formulation Yeast extract 0.5g/L, Proteose Peptone No. 3 0.5g/L, Casamino Acids 0.5g/L, Dextrose 0.5g/L, Soluble Starch 0.5g/L, Sodium Pyruvate 0.3g/L, Dipotassium Phosphate 0.3g/L, Magnesium Sulfate 0.05g/L, Agar 15g/L
For Use With (Application) Low nutrient medium for enumeration and cultivation of bacteria from potable Water
Packaging Type Bottle
Quantity 500 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.