Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD Difco™ Salmonella O Antisera and Quality Control Antigens

Catalog No. B228191 Shop All BD Diagnostic Systems Products
Change view
Click to view available options
Product Type:
Salmonella O Antiserum Factor 10, Group E1
Salmonella O Antiserum Factor 14
Salmonella O Antiserum Factor 15, Groups E2 and E3
Salmonella O Antiserum Factor 19, Group E4
Salmonella O Antiserum Factor 2
Salmonella O Antiserum Factor 20
Salmonella O Antiserum Factor 22, Group G1
Salmonella O Antiserum Factor 23, Group G2
Salmonella O Antiserum Factor 34, Group E3
Salmonella O Antiserum Factor 4, Group B
Salmonella O Antiserum Factor 5, Group B
Salmonella O Antiserum Factor 7, Groups C1 and C4
Salmonella O Antiserum Factor 8, Groups C2 and C3
Salmonella O Antiserum Factor 9, Group D
Salmonella O Antiserum Factors 4, 5, Group B
Salmonella O Antiserum Group D1 Factors 1, 9, 12
Salmonella O Antiserum Group E, Factors 1, 3, 10, 15, 19, 34
Salmonella O Antiserum Group E1 Factors 3, 10
Salmonella O Antiserum Group F Factor 11
Salmonella O Antiserum Group G Factors 13, 22
Salmonella O Antiserum Group H Factors 1, 6, 14, 24, 25
Salmonella O Antiserum Group J Factor 17
Salmonella O Antiserum Group M Factor 28
Salmonella O Antiserum Poly A, Groups A, B, D, E1, E2, E3, E4 and L
Salmonella O Antiserum Poly B, Groups C1, C2, F, G and H
Salmonella O Antiserum Poly C, Groups I, J, K, M, N and O
Salmonella O Antiserum Poly D, Groups P, Q, R, S, T and U
Salmonella O Antiserum Poly E, Groups V, W, X, Y and Z
Salmonella O Antiserum Poly F, Groups 51-55
Salmonella O Antiserum Poly G, Groups 56-61
30 product options available for selection
Product selection table with 30 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type
B228191 Salmonella O Antiserum Group E, Factors 1, 3, 10, 15, 19, 34
DF2517-47-4 Salmonella O Antiserum Group J Factor 17
B222611 Salmonella O Antiserum Group G Factors 13, 22
B222621 Salmonella O Antiserum Group H Factors 1, 6, 14, 24, 25
D266147 Salmonella O Antiserum Factor 14
B229511 Salmonella O Antiserum Group D1 Factors 1, 9, 12
DF2257-47-1 Salmonella O Antiserum Factor 10, Group E1
DF2258-47-0 Salmonella O Antiserum Factor 15, Groups E2 and E3
DF2259-47-9 Salmonella O Antiserum Factor 19, Group E4
DF2260-47-6 Salmonella O Antiserum Group F Factor 11
DF2512-47-2 Salmonella O Antiserum Factor 34, Group E3
DF2535-47-5 Salmonella O Antiserum Poly B, Groups C1, C2, F, G and H
DF2534-47-6 Salmonella O Antiserum Poly A, Groups A, B, D, E1, E2, E3, E4 and L
DF2536-47-4 Salmonella O Antiserum Poly C, Groups I, J, K, M, N and O
DF2537-47-3 Salmonella O Antiserum Poly D, Groups P, Q, R, S, T and U
DF2538-47-2 Salmonella O Antiserum Poly E, Groups V, W, X, Y and Z
DF2645-47-2 Salmonella O Antiserum Poly F, Groups 51-55
DF2662-47-0 Salmonella O Antiserum Factor 20
DF2659-47-5 Salmonella O Antiserum Factor 4, Group B
DF2646-47-1 Salmonella O Antiserum Poly G, Groups 56-61
DF2660-47-2 Salmonella O Antiserum Factor 5, Group B
DF2664-47-8 Salmonella O Antiserum Factor 23, Group G2
DF2663-47-9 Salmonella O Antiserum Factor 22, Group G1
DF2814-47-7 Salmonella O Antiserum Factor 2
DF2815-47-6 Salmonella O Antiserum Factors 4, 5, Group B
DF2816-47-5 Salmonella O Antiserum Factor 7, Groups C1 and C4
DF2817-47-4 Salmonella O Antiserum Factor 8, Groups C2 and C3
DF2818-47-3 Salmonella O Antiserum Factor 9, Group D
DF2952-47-9 Salmonella O Antiserum Group E1 Factors 3, 10
DF2520-47-2 Salmonella O Antiserum Group M Factor 28
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 30 options available.
30 options
This item is not returnable. View return policy
The products indicated contain mercury. Don't put products containing mercury in the trash. Products containing mercury are to be recycled or disposed of as hazardous waste. Products containing mercury may be regulated by Local, State, or Federal Laws. Some products containing mercury are not available for sale in some states.
Catalog No. B228191 Supplier BD Supplier No. 228191
Add to Cart
Add to Cart
This item is not returnable. View return policy
The products indicated contain mercury. Don't put products containing mercury in the trash. Products containing mercury are to be recycled or disposed of as hazardous waste. Products containing mercury may be regulated by Local, State, or Federal Laws. Some products containing mercury are not available for sale in some states.

Salmonella species cause a variety of human diseases called salmonelloses. The range of disease is from mild self-limiting gastroenteritis to more severe forms, possibly with bacteremia or typhoid fever, which can be life threatening. Hence diagnosis and identification of correct serotype is essential. These antisera aid in detecting such serotypes and the quality control antigens check their performance.

  • Lyophilized, polyclonal rabbit antisera
  • Polyvalent, specific for somatic antigens
  • Cross-reactions are possible because serogroups may share non-major group antigens
  • When properly rehydrated and used as recommended, each vial contains sufficient reagent for 60 tests
  • For in vitro diagnostic use
  • containing approximately 0.2% sodium azide as a preservative

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance For in vitro diagnostic use
Product Type Salmonella O Antiserum Group E, Factors 1, 3, 10, 15, 19, 34
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.