Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD Difco™ Salmonella O QC Antigens

Catalog No. DF2134504 Shop All Waters Corporation Products
Change view
Click to view available options
Product Type:
Salmonella O Group A Antigen
Salmonella O Group D Antigen
Salmonella O Group E1 Antigen
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type
DF2134-50-4 Salmonella O Group D Antigen
DF2130-50-8 Salmonella O Group A Antigen
DF2135-50-3 Salmonella O Group E1 Antigen
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
The products indicated contain mercury. Don't put products containing mercury in the trash. Products containing mercury are to be recycled or disposed of as hazardous waste. Products containing mercury may be regulated by Local, State, or Federal Laws. Some products containing mercury are not available for sale in some states.
Catalog No. DF2134-50-4 Supplier BD Supplier No. 221341
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
The products indicated contain mercury. Don't put products containing mercury in the trash. Products containing mercury are to be recycled or disposed of as hazardous waste. Products containing mercury may be regulated by Local, State, or Federal Laws. Some products containing mercury are not available for sale in some states.

Quick reactive Gold standard antisera-typing panels for Salmonella O antigen

  • Product type: Antigens and Antisera
  • Groupings: Q through S products
  • Sub type: Quality control antigens
  • Type: Antigens
  • Volume: 1mL
  • Shelf life: 730 days

Certifications

CE marked

TRUSTED_SUSTAINABILITY

Specifications

Product Type Salmonella O Group D Antigen
For Use With (Application) Used in the quality control of Salmonella O Antisera Poly B (225351), Factor 7 (228161), Group C1 Factors 6 (240985), and Poly A-I & Vi (240993)
Quantity 1 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.