Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
BD FoxP3 Staining Kit
High performance reagent kit for detection of human FoxP3 positive regulatory T cells
Supplier: BD 560132
Includes: PE/Alexa Fluor™ 647/ Alexa Fluor 488, FoxP3, CD4, CD25, Buffer set
Description
- Complementing antibody conjugates buffer system fixes and permeabilizes cells in easy, simple steps
- Components are sufficient for 100 tests
- Clone: M-A251, RPA-T4, 259D/C7
- Isotype: mouse IgG1, IgG1, κ, IgG1, κ
Flow Cytometry

Specifications
Alexa Fluor 647 | |
1 Kit |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Research use only
Spot an opportunity for improvement?Share a Content Correction