Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD FUT-175 (Futhan)

Catalog No. BDB552035 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB552035 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB552035 Supplier BD Supplier No. 552035
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Synthetic, broad-specificity protease inhibitor and an inhibitor of the classical and alternate pathways of complement activation

  • Alias: Futhan
  • Chemical name: 6-amidino-2-naphthyl p-guanidinobenzoate dimethanesulfonate
  • Molecular weight: 539.58 Da
  • Storage buffer: Lyophilized powder

Serum Stab

TRUSTED_SUSTAINABILITY

Specifications

For Use With (Application) Serum Stab
Content And Storage 4°C
Quantity 5 mg
Molecular Weight (g/mol) 539.58
Inhibitors FUT-175 (Futhan)
Form Lyophilized Powder

Research use only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.