Learn More
Description
- Format: PE
- Reactivity: Ms
- Isotype: Mouse IgG1, Λ
Specifications
Specifications
| Concentration | 0.5mg/mL |
| For Use With (Application) | ELISA, Flow Cytometry |
| Formulation | Aqueous Buffered solution containing 0.09% Sodium Azide |
| Name | Mouse PE h-2Kb:ig bd dimerX Recombinant Fusion Protein |
| pH Range | 7.2 |
| Purification Method | Affinity Chromatography, SDS-PAGE |
| Quantity | 0.1 mg |
| Storage Requirements | 4°C |
| Recombinant | Recombinant |
| Form | Solution |
Safety and Handling
Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.