Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD Human Mesenchymal Stem Cell Analysis Kit

Catalog No. BDB562245 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB562245 50 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB562245 Supplier BD Supplier No. 562245
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Includes

  • PE hMSC Negative Cocktail
  • PE hMSC Isotype Control Negative Cocktail
  • hMSC Positive Cocktail
  • hMSC Isotype Control Positive Cocktail
  • FITC Mouse Anti-Human CD90
  • PE Mouse Anti-Human CD44
  • PE Mouse IgG2b, k Isotype Contro
  • APC Mouse Anti-Human CD73
  • PerCP-Cy5.5 Mouse Anti-Human CD105 (Endoglin)

For reproducible multicolor phenotyping of MSC expansions

Provides comprehensive system for the reliable, in-depth characterization of human bone marrow-derived multipotent mesenchymal stromal cells (BM-MSCs) based on the phenotypic signature described by the The Mesenchymal and Tissue Stem Cell Committee of the International Society for Cellular Therapy (ISCT).

  • Modular designed for additional markers to be “dropped in”
  • Contains pre-conjugated and pre-titrated cocktails of the ISCT-defined positive expression markers (CD105 PerCP-Cy5.5/CD73 APC/CD90 FITC) and negative expression marker
  • Enables the easy addition of supplementary monoclonal antibodies against critical cell surface marker

TRUSTED_SUSTAINABILITY

Specifications

Quantity 50 Tests
Content And Storage 4°C
For Use With (Application) For reproducible multicolor phenotyping of MSC expansions, FCM
Reactivity Human
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.